Assay Details
| Gene Symbol: BIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105572.5 | NP_177064.1 | 5 - 6 | 891 | 75 | |
| GenBank mRNA | BT002352.1 | - | 5 - 6 | 716 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BIM2| Gene Symbol: BIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105572.5 | NP_177064.1 | 5 - 6 | 891 | 75 | |
| GenBank mRNA | BT002352.1 | - | 5 - 6 | 716 | 75 | |
Set Membership: |
|