Assay Details
| Gene Symbol: AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334400.1 | NP_001320362.1 | 5 - 6 | 720 | 119 | |
| NM_001334401.1 | NP_001320361.1 | 5 - 6 | 474 | 119 | ||
| NM_105581.3 | NP_177074.1 | 5 - 6 | 720 | 119 | ||
| GenBank mRNA | BT004113.1 | - | 5 - 6 | 615 | 119 | |
| BT004951.1 | - | 5 - 6 | 473 | 119 | ||
More Information
Set Membership: |
|

AP1
Genomic Map