Assay Details
| Gene Symbol: CRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105585.3 | NP_177078.1 | 3 - 4 | 533 | 74 | |
| GenBank mRNA | AF132606.1 | - | 3 - 4 | 356 | 74 | |
| AK229609.1 | - | 3 - 4 | 411 | 74 | ||
| BT008618.1 | - | 3 - 4 | 411 | 74 | ||
| DQ446412.1 | - | 3 - 4 | 332 | 74 | ||
More Information
Set Membership: |
|

CRC
Genomic Map