Assay Details
| Gene Symbol: FLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105587.6 | NP_177080.3 | 2 - 3 | 382 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLN2| Gene Symbol: FLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105587.6 | NP_177080.3 | 2 - 3 | 382 | 105 | |
Set Membership: |
|