Assay Details
| Gene Symbol: CM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105604.2 | NP_177096.1 | 3 - 4 | 619 | 62 | |
| GenBank mRNA | AK117860.1 | - | 3 - 4 | 493 | 62 | |
| BT005306.1 | - | 3 - 4 | 477 | 62 | ||
More Information
Set Membership: |
|

CM3
Genomic Map