Assay Details
Gene Symbol: AGO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105611.5 | NP_177103.1 | 2 - 3 | 1898 | 145 | |
GenBank mRNA | AY394564.1 | - | 2 - 3 | 1736 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
