Assay Details
| Gene Symbol: NAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105616.4 | NP_564966.1 | 1 - 2 | 317 | 103 | |
| GenBank mRNA | AF360201.1 | - | 1 - 2 | 257 | 103 | |
| AY051015.1 | - | 1 - 2 | 164 | 103 | ||
More Information
Set Membership: |
|

NAP
Genomic Map