Assay Details
| Gene Symbol: NAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105616.4 | NP_564966.1 | 2 - 3 | 601 | 92 | |
| GenBank mRNA | AF360201.1 | - | 2 - 3 | 541 | 92 | |
| AY051015.1 | - | 2 - 3 | 448 | 92 | ||
More Information
Set Membership: |
|

NAP
Genomic Map