Assay Details
| Gene Symbol: AT1G69660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105634.1 | NP_177124.1 | 3 - 4 | 544 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G69660| Gene Symbol: AT1G69660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105634.1 | NP_177124.1 | 3 - 4 | 544 | 65 | |
Set Membership: |
|