Assay Details
| Gene Symbol: HO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124102.1 | NP_001117574.1 | 2 - 3 | 529 | 76 | |
| NM_001334420.1 | NP_001321799.1 | 1 - 2 | 525 | 76 | ||
| NM_105640.4 | NP_177130.1 | 2 - 3 | 664 | 76 | ||
| GenBank mRNA | AY084250.1 | - | 2 - 3 | 529 | 76 | |
More Information
Set Membership: |
|

HO3
Genomic Map