Assay Details
| Gene Symbol: HO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334420.1 | NP_001321799.1 | 2 - 3 | 743 | 142 | |
| NM_105640.4 | NP_177130.1 | 3 - 4 | 882 | 142 | ||
| GenBank mRNA | AY084250.1 | - | 3 - 4 | 747 | 142 | |
More Information
Set Membership: |
|

HO3
Genomic Map