Assay Details
| Gene Symbol: AT1G69760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105644.3 | NP_564974.1 | 1 | 1377 | 166 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G69760| Gene Symbol: AT1G69760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105644.3 | NP_564974.1 | 1 | 1377 | 166 | |
Set Membership: |
|