Assay Details
| Gene Symbol: AMY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105651.5 | NP_564977.1 | 4 - 5 | 756 | 149 | |
| GenBank mRNA | AY050398.1 | - | 4 - 5 | 632 | 149 | |
| BT000643.1 | - | 4 - 5 | 586 | 149 | ||
More Information
Set Membership: |
|

AMY3
Genomic Map