Assay Details
| Gene Symbol: NRT1:2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105653.5 | NP_564978.1 | 4 - 5 | 1027 | 139 | |
| GenBank mRNA | BT000720.1 | - | 4 - 5 | 782 | 139 | |
More Information
Set Membership: |
|

NRT1:2
Genomic Map