Assay Details
| Gene Symbol: ABCB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105729.6 | NP_177218.3 | 2 - 3 | 767 | 76 | |
| GenBank mRNA | AK228750.1 | - | 2 - 3 | 705 | 76 | |
| AY078409.1 | - | 2 - 3 | 580 | 76 | ||
More Information
Set Membership: |
|

ABCB26
Genomic Map