Assay Details
| Gene Symbol: ABCB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334463.1 | NP_001321943.1 | 5 - 6 | 926 | 136 | |
| NM_105729.6 | NP_177218.3 | 5 - 6 | 975 | 136 | ||
| GenBank mRNA | AK228750.1 | - | 5 - 6 | 913 | 136 | |
| AY078409.1 | - | 5 - 6 | 788 | 136 | ||
More Information
Set Membership: |
|

ABCB26
Genomic Map