Assay Details
| Gene Symbol: TIFY7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036187.5 | NP_001031264.4 | 1 - 2 | 273 | 100 | |
| NM_001334477.1 | NP_001320988.1 | 1 - 2 | 273 | 100 | ||
| NM_105738.8 | NP_177227.5 | 1 - 2 | 512 | 100 | ||
More Information
Set Membership: |
|

TIFY7
Genomic Map