Assay Details
| Gene Symbol: TIFY7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036187.5 | NP_001031264.4 | 3 - 4 | 507 | 71 | |
| NM_001334477.1 | NP_001320988.1 | 3 - 4 | 507 | 71 | ||
| NM_105738.8 | NP_177227.5 | 3 - 4 | 746 | 71 | ||
More Information
Set Membership: |
|

TIFY7
Genomic Map