Assay Details
| Gene Symbol: HB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334482.1 | NP_001321391.1 | 2 - 3 | 678 | 100 | |
| NM_105760.6 | NP_177248.3 | 3 - 4 | 590 | 100 | ||
| GenBank mRNA | AK175837.1 | - | 3 - 4 | 447 | 100 | |
| AK175965.1 | - | 3 - 4 | 447 | 100 | ||
| EU339186.1 | - | 3 - 4 | 528 | 100 | ||
More Information
Set Membership: |
|

HB18
Genomic Map