Assay Details
| Gene Symbol: AT1G70950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105763.4 | NP_177251.2 | 3 - 4 | 1015 | 131 | |
| GenBank mRNA | AK176599.1 | - | 3 - 4 | 913 | 131 | |
More Information
Set Membership: |
|

AT1G70950
Genomic Map