Assay Details
| Gene Symbol: MYBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334485.1 | NP_001321410.1 | 1 - 2 | 636 | 67 | |
| NM_105772.3 | NP_177259.1 | 2 - 3 | 463 | 67 | ||
| GenBank mRNA | AF370609.1 | - | 2 - 3 | 160 | 67 | |
| AY048231.1 | - | 2 - 3 | 220 | 67 | ||
| AY087093.1 | - | 2 - 3 | 296 | 67 | ||
| AY091697.1 | - | 2 - 3 | 160 | 67 | ||
| AY519519.1 | - | 2 - 3 | 160 | 67 | ||
More Information
Set Membership: |
|

MYBL2
Genomic Map