Assay Details
| Gene Symbol: SAG18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105788.4 | NP_177275.1 | 2 - 3 | 312 | 97 | |
| GenBank mRNA | BT044602.1 | - | 2 - 3 | 234 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAG18| Gene Symbol: SAG18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105788.4 | NP_177275.1 | 2 - 3 | 312 | 97 | |
| GenBank mRNA | BT044602.1 | - | 2 - 3 | 234 | 97 | |
Set Membership: |
|