Assay Details
| Gene Symbol: SAG18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198440.1 | NP_001185369.1 | 4 - 5 | 500 | 110 | |
| NM_105788.4 | NP_177275.1 | 5 - 6 | 673 | 110 | ||
| GenBank mRNA | BT044602.1 | - | 5 - 6 | 595 | 110 | |
More Information
Set Membership: |
|

SAG18
Genomic Map