Assay Details
| Gene Symbol: AT1G71350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105804.4 | NP_177291.2 | 5 - 6 | 1099 | 97 | |
| GenBank mRNA | AK227085.1 | - | 5 - 6 | 1048 | 97 | |
More Information
Set Membership: |
|

AT1G71350
Genomic Map