Assay Details
| Gene Symbol: AT1G71360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334496.1 | NP_001320414.1 | 2 | 1764 | 63 | |
| NM_105805.6 | NP_177292.4 | 3 | 2146 | 63 | ||
More Information
Set Membership: |
|

AT1G71360
Genomic Map