Assay Details
| Gene Symbol: PFI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105813.4 | NP_565017.1 | 9 - 10 | 1211 | 87 | |
| GenBank mRNA | AK118835.1 | - | 9 - 10 | 1164 | 87 | |
| BT002403.1 | - | 9 - 10 | 1166 | 87 | ||
| BT008888.1 | - | 9 - 10 | 1153 | 87 | ||
More Information
Set Membership: |
|

PFI
Genomic Map