Assay Details
| Gene Symbol: CK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105828.4 | NP_177315.1 | 1 - 2 | 933 | 95 | |
| GenBank mRNA | AK229787.1 | - | 1 - 2 | 571 | 95 | |
| AK230407.1 | - | 1 - 2 | 570 | 95 | ||
| BT008311.1 | - | 1 - 2 | 364 | 95 | ||
More Information
Set Membership: |
|

CK1
Genomic Map