Assay Details
| Gene Symbol: PAB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198444.1 | NP_001185373.1 | 4 - 5 | 1446 | 98 | |
| NM_105835.2 | NP_177322.2 | 4 - 5 | 1464 | 98 | ||
More Information
Set Membership: |
|

PAB5
Genomic Map