Assay Details
| Gene Symbol: CPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105837.4 | NP_177324.2 | 1 - 2 | 193 | 112 | |
| GenBank mRNA | BT029459.1 | - | 1 - 2 | 129 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPB| Gene Symbol: CPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105837.4 | NP_177324.2 | 1 - 2 | 193 | 112 | |
| GenBank mRNA | BT029459.1 | - | 1 - 2 | 129 | 112 | |
Set Membership: |
|