Assay Details
| Gene Symbol: PUM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334523.1 | NP_001321803.1 | 10 - 11 | 2197 | 116 | |
| NM_105891.3 | NP_177376.1 | 10 - 11 | 2047 | 116 | ||
| NM_179547.5 | NP_849878.2 | 10 - 11 | 1982 | 116 | ||
| NM_202401.1 | NP_974130.1 | 10 - 11 | 1952 | 116 | ||
| GenBank mRNA | AY035041.1 | - | 10 - 11 | 1989 | 116 | |
| AY051073.1 | - | 9 - 10 | 1878 | 116 | ||
More Information
Set Membership: |
|

PUM23
Genomic Map