Assay Details
| Gene Symbol: PUM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334523.1 | NP_001321803.1 | 3 - 4 | 932 | 117 | |
| NM_105891.3 | NP_177376.1 | 3 - 4 | 782 | 117 | ||
| NM_179547.5 | NP_849878.2 | 3 - 4 | 717 | 117 | ||
| NM_202401.1 | NP_974130.1 | 3 - 4 | 687 | 117 | ||
| GenBank mRNA | AY035041.1 | - | 3 - 4 | 724 | 117 | |
| AY051073.1 | - | 2 - 3 | 613 | 117 | ||
More Information
Set Membership: |
|

PUM23
Genomic Map