Assay Details
| Gene Symbol: PUM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334523.1 | NP_001321803.1 | 7 - 8 | 1673 | 79 | |
| NM_105891.3 | NP_177376.1 | 7 - 8 | 1523 | 79 | ||
| NM_179547.5 | NP_849878.2 | 7 - 8 | 1458 | 79 | ||
| NM_202401.1 | NP_974130.1 | 7 - 8 | 1428 | 79 | ||
| GenBank mRNA | AY035041.1 | - | 7 - 8 | 1465 | 79 | |
| AY051073.1 | - | 6 - 7 | 1354 | 79 | ||
More Information
Set Membership: |
|

PUM23
Genomic Map