Assay Details
| Gene Symbol: ERF73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084343.2 | NP_001077812.1 | 2 | 977 | 82 | |
| NM_001124115.1 | NP_001117587.1 | 2 | 860 | 82 | ||
| NM_105895.2 | NP_177380.2 | 1 | 946 | 82 | ||
| GenBank mRNA | BT002063.1 | - | 1 | 946 | 82 | |
More Information
Set Membership: |
|

ERF73
Genomic Map