Assay Details
| Gene Symbol: EDA25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105903.4 | NP_177388.2 | 12 - 13 | 2115 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDA25| Gene Symbol: EDA25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105903.4 | NP_177388.2 | 12 - 13 | 2115 | 68 | |
Set Membership: |
|