Assay Details
| Gene Symbol: JAZ6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334533.1 | NP_001321693.1 | 1 - 2 | 342 | 74 | |
| NM_105904.4 | NP_565043.1 | 1 - 2 | 587 | 74 | ||
| GenBank mRNA | AF370147.1 | - | 1 - 2 | 258 | 74 | |
| AY059089.1 | - | 1 - 2 | 161 | 74 | ||
| AY088035.1 | - | 1 - 2 | 313 | 74 | ||
More Information
Set Membership: |
|

JAZ6
Genomic Map