Assay Details
| Gene Symbol: LOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105911.5 | NP_177396.1 | 2 - 3 | 789 | 71 | |
| GenBank mRNA | AJ302042.1 | - | 2 - 3 | 691 | 71 | |
| AY056166.1 | - | 2 - 3 | 783 | 71 | ||
| AY091193.1 | - | 2 - 3 | 691 | 71 | ||
More Information
Set Membership: |
|

LOX4
Genomic Map