Assay Details
| Gene Symbol: TRFL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105924.4 | NP_565045.1 | 6 - 7 | 1796 | 71 | |
| NM_202404.2 | NP_974133.1 | 6 - 7 | 1814 | 71 | ||
| GenBank mRNA | AY045628.1 | - | 6 - 7 | 1722 | 71 | |
| AY124868.1 | - | 5 - 6 | 1453 | 71 | ||
| AY519540.1 | - | 5 - 6 | 1453 | 71 | ||
More Information
Set Membership: |
|

TRFL6
Genomic Map