Assay Details
| Gene Symbol: TRFL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105924.4 | NP_565045.1 | 8 - 9 | 2132 | 58 | |
| NM_202404.2 | NP_974133.1 | 8 - 9 | 2150 | 58 | ||
| GenBank mRNA | AY045628.1 | - | 8 - 9 | 2058 | 58 | |
| AY124868.1 | - | 7 - 8 | 1789 | 58 | ||
| AY519540.1 | - | 7 - 8 | 1789 | 58 | ||
More Information
Set Membership: |
|

TRFL6
Genomic Map