Assay Details
| Gene Symbol: CAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105927.4 | NP_177412.1 | 5 - 6 | 1020 | 102 | |
| GenBank mRNA | AY288079.1 | - | 5 - 6 | 872 | 102 | |
| BT002440.1 | - | 5 - 6 | 972 | 102 | ||
| BT008840.1 | - | 5 - 6 | 872 | 102 | ||
More Information
Set Membership: |
|

CAD1
Genomic Map