Assay Details
| Gene Symbol: HAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084346.3 | NP_001077815.1 | 2 - 3 | 1405 | 89 | |
| NM_001198456.1 | NP_001185385.1 | 2 - 3 | 1331 | 89 | ||
| NM_001334550.1 | NP_001320804.1 | 2 - 3 | 1352 | 89 | ||
| NM_001334551.1 | NP_001320805.1 | 2 - 3 | 1345 | 89 | ||
| NM_105936.4 | NP_177421.1 | 2 - 3 | 1405 | 89 | ||
| GenBank mRNA | AK230171.1 | - | 2629 | 89 | ||
| BT015409.1 | - | 1 - 2 | 746 | 89 | ||
More Information
Set Membership: |
|

HAB1
Genomic Map