Assay Details
| Gene Symbol: BGAL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036196.2 | NP_001031273.1 | 11 - 12 | 1493 | 86 | |
| NM_001334559.1 | NP_001321344.1 | 11 - 12 | 1488 | 86 | ||
| NM_001334560.1 | NP_001321343.1 | 12 - 13 | 1345 | 86 | ||
| NM_105957.3 | NP_565051.1 | 11 - 12 | 1480 | 86 | ||
| GenBank mRNA | AY058198.1 | - | 11 - 12 | 1382 | 86 | |
| AY142008.1 | - | 11 - 12 | 1161 | 86 | ||
More Information
Set Membership: |
|

BGAL17
Genomic Map