Assay Details
| Gene Symbol: BGAL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036196.2 | NP_001031273.1 | 15 - 16 | 1958 | 100 | |
| NM_001334559.1 | NP_001321344.1 | 15 - 16 | 1953 | 100 | ||
| NM_001334560.1 | NP_001321343.1 | 16 - 17 | 1810 | 100 | ||
| NM_105957.3 | NP_565051.1 | 15 - 16 | 1945 | 100 | ||
| GenBank mRNA | AY058198.1 | - | 15 - 16 | 1847 | 100 | |
| AY142008.1 | - | 15 - 16 | 1626 | 100 | ||
More Information
Set Membership: |
|

BGAL17
Genomic Map