Assay Details
| Gene Symbol: BGAL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036196.2 | NP_001031273.1 | 9 - 10 | 1350 | 101 | |
| NM_001334559.1 | NP_001321344.1 | 9 - 10 | 1345 | 101 | ||
| NM_001334560.1 | NP_001321343.1 | 10 - 11 | 1202 | 101 | ||
| NM_105957.3 | NP_565051.1 | 9 - 10 | 1337 | 101 | ||
| GenBank mRNA | AY058198.1 | - | 9 - 10 | 1239 | 101 | |
| AY142008.1 | - | 9 - 10 | 1018 | 101 | ||
More Information
Set Membership: |
|

BGAL17
Genomic Map