Assay Details
| Gene Symbol: BGLU42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036898.2 | NP_001031975.1 | 4 - 5 | 446 | 160 | |
| NM_123047.4 | NP_198505.2 | 4 - 5 | 447 | 160 | ||
| GenBank mRNA | AK175760.1 | - | 4 - 5 | 447 | 160 | |
| BT010611.1 | - | 4 - 5 | 307 | 160 | ||
More Information
Set Membership: |
|

BGLU42
Genomic Map