Assay Details
| Gene Symbol: CAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123052.5 | NP_198510.2 | 10 - 11 | 1465 | 117 | |
| GenBank mRNA | AK117747.1 | - | 10 - 11 | 1270 | 117 | |
More Information
Set Membership: |
|

CAT3
Genomic Map