Assay Details
| Gene Symbol: CAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123052.5 | NP_198510.2 | 4 - 5 | 769 | 126 | |
| GenBank mRNA | AK117747.1 | - | 4 - 5 | 574 | 126 | |
More Information
Set Membership: |
|

CAT3
Genomic Map