Assay Details
| Gene Symbol: CAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123052.5 | NP_198510.2 | 6 - 7 | 978 | 91 | |
| GenBank mRNA | AK117747.1 | - | 6 - 7 | 783 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAT3| Gene Symbol: CAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123052.5 | NP_198510.2 | 6 - 7 | 978 | 91 | |
| GenBank mRNA | AK117747.1 | - | 6 - 7 | 783 | 91 | |
Set Membership: |
|