Assay Details
| Gene Symbol: AT5G37420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123101.1 | NP_198558.1 | 1 - 2 | 1205 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G37420| Gene Symbol: AT5G37420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123101.1 | NP_198558.1 | 1 - 2 | 1205 | 99 | |
Set Membership: |
|