Assay Details
| Gene Symbol: AT5G37490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123108.4 | NP_198565.1 | 1 | 1413 | 60 | |
| GenBank mRNA | BT020311.1 | - | 1 | 1321 | 60 | |
| BT020439.1 | - | 1 | 1098 | 60 | ||
More Information
Set Membership: |
|

AT5G37490
Genomic Map