Assay Details
| Gene Symbol: ALY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085207.2 | NP_001078676.1 | 2 - 3 | 494 | 88 | |
| NM_123131.4 | NP_198588.2 | 2 - 3 | 416 | 88 | ||
| GenBank mRNA | AK227931.1 | - | 2 - 3 | 366 | 88 | |
| BT010588.1 | - | 1 - 2 | 189 | 88 | ||
More Information
Set Membership: |
|

ALY4
Genomic Map